Learn More
Imeb Inc Small Pathology Bone Band Saw Optional Diamond Blade, Optional Cooling System is Required for Diamond Blades, Bone Band Saw Sold Separately
Supplier: Imeb Inc SD311
Saw Blade Diamond is .125" wide X .025" for IMEB bone band saw that is sold separately. Optional Cooling System is required for diamond blades. The IMEB, Inc. Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw is only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.