CerCo Diamonite™ Mortar and Pestle Sets DFS Item
Synthetic sapphire mortars offer excellent hardness and chemical resistance
Supplier:  CerCo DPMMP100
 View more versions of this product
Catalog No. 12-952-5
Explore available promotions
Alert:
Not recommended for cryogenic use.
Description
Description
Synthetic sapphire mortars offer excellent hardness and chemical resistance
  • Made of finely powdered synthetic sapphire
  • Molded under extremely high pressure and sintered at high temperature
  • Hard as tungsten carbide but less costly; hardness is 9+ on Mohs scale
  • Resistant to acids, alkalis, corrosive chemicals
  • Grinds materials not easily reduced in porcelain or metal mortars
Specifications
Specifications
3.9 in.
10cm
Mortar and Pestle
For grinding or crushing
5cm
3.9 in.
4.8 in.
12.2cm
Diamonite
2 in.
10cm