Learn More
Diatrizoic acid, Thermo Scientific Chemicals
A high-osmolality radiocontrast agent
$130.74 - $383.49
Chemical Identifiers
CAS | 117-96-4 |
---|---|
Molecular Formula | C11H9I3N2O4 |
Molecular Weight (g/mol) | 613.92 |
MDL Number | MFCD00069960 |
InChI Key | YVPYQUNUQOZFHG-UHFFFAOYSA-N |
Synonym | diatrizoic acid, amidotrizoic acid, diatrizoate, amidotrizoate, urogranoic acid, urografin acid, odiston, urotrast, diat, diatriazoate |
PubChem CID | 2140 |
ChEBI | CHEBI:53691 |
IUPAC Name | 3,5-diacetamido-2,4,6-triiodobenzoic acid |
SMILES | CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAJ6344414
|
Thermo Scientific Chemicals
J6344414 |
25 g |
Each for $130.74
|
|
||||||
AAJ6344422
|
Thermo Scientific Chemicals
J6344422 |
100 g |
Each for $383.49
|
|
||||||
Description
Diatrizoic acid is a gradient reagent used for blood cell separation and diagnostic aid (radiopaque medium). Diatrizoic acid may be used as an alternative to barium sulfate for medical imaging of the gastrointestinal tract, such as upper gastrointestinal series and small bowel series.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in Methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.
Chemical Identifiers
117-96-4 | |
613.92 | |
YVPYQUNUQOZFHG-UHFFFAOYSA-N | |
2140 | |
3,5-diacetamido-2,4,6-triiodobenzoic acid |
C11H9I3N2O4 | |
MFCD00069960 | |
diatrizoic acid, amidotrizoic acid, diatrizoate, amidotrizoate, urogranoic acid, urografin acid, odiston, urotrast, diat, diatriazoate | |
CHEBI:53691 | |
CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I |
Specifications
117-96-4 | |
C11H9I3N2O4 | |
25 g | |
diatrizoic acid, amidotrizoic acid, diatrizoate, amidotrizoate, urogranoic acid, urografin acid, odiston, urotrast, diat, diatriazoate | |
YVPYQUNUQOZFHG-UHFFFAOYSA-N | |
3,5-diacetamido-2,4,6-triiodobenzoic acid | |
2140 | |
613.9 | |
Diatrizoic acid |
White | |
MFCD00069960 | |
14,2992 | |
Soluble in Methanol. | |
CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I | |
613.92 | |
CHEBI:53691 | |
Powder |
Safety and Handling
EINECSNumber : 204-223-6
RTECSNumber : DG5950000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only