missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzo[a,e]cyclooctene 97.0+%, TCI America™
$242.33 - $820.19
Chemical Identifiers
| CAS | 262-89-5 |
|---|---|
| Molecular Formula | C16H12 |
| Molecular Weight (g/mol) | 204.272 |
| InChI Key | VGQWCQNHAMVTJY-YAZGSDAZSA-N |
| Synonym | Dibenzo[a,e]cyclooctatetraene, Dibenzo[a,e][8]annulene, DBCOT |
| PubChem CID | 6439814 |
| IUPAC Name | (4aZ,6Z,10aZ,12Z)-dibenzo[2,1-a |
| SMILES | C1=CC2=CC=C3C=CC=CC3=CC=C2C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4338200MG
|
TCI America
D4338200MG |
200 mg |
Each for $242.33
|
|
|||||
|
D43381G
|
TCI America
D43381G |
1 g |
Each for $820.19
|
|
|||||
Chemical Identifiers
| 262-89-5 | |
| 204.272 | |
| Dibenzo[a,e]cyclooctatetraene, Dibenzo[a,e][8]annulene, DBCOT | |
| (4aZ,6Z,10aZ,12Z)-dibenzo[2,1-a |
| C16H12 | |
| VGQWCQNHAMVTJY-YAZGSDAZSA-N | |
| 6439814 | |
| C1=CC2=CC=C3C=CC=CC3=CC=C2C=C1 |
Specifications
| 262-89-5 | |
| White-Yellow | |
| C16H12 | |
| Dibenzo[a,e]cyclooctatetraene, Dibenzo[a,e][8]annulene, DBCOT | |
| C1=CC2=CC=C3C=CC=CC3=CC=C2C=C1 | |
| 204.272 | |
| 204.27 | |
| Crystalline Powder |
| 107°C | |
| 130°C | |
| 200 mg | |
| VGQWCQNHAMVTJY-YAZGSDAZSA-N | |
| (4aZ,6Z,10aZ,12Z)-dibenzo[2,1-a | |
| 6439814 | |
| ≥97.0% (GC) | |
| Dibenzo[a,e]cyclooctene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator