Learn More
Dibenzosuberone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 114741000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Dibenzosuberone | |
1210-35-1 | |
Yellow | |
148°C (0.3 mmHg) | |
96% min. (GC) | |
C15H12O | |
MFCD00003587 | |
1.15 | |
Solubility in water: insoluble | |
O=C1C2=CC=CC=C2CCC2=CC=CC=C12 | |
208.26 | |
208.26 | |
Liquid or Low Melting Solid |
97% | |
32°C to 34°C | |
1.1500g/mL | |
Authentic | |
Glass bottle | |
1.6332 to 1.6352 | |
100 g | |
dibenzosuberone, dibenzocycloheptenone, dibenzosuberan-5-one, dibenzsuberone, 5-dibenzosuberone, 2,3:6,7-dibenzosuberone, 5h-dibenzo a,d cyclohepten-5-one, 10,11-dihydro, unii-8etk71th0h, dibenzo b,f cycloheptan-1-one, ccris 2780 | |
BMVWCPGVLSILMU-UHFFFAOYSA-N | |
5,6-dihydrodibenzo[2,1-b | |
14589 | |
97% |
Identifiants chimiques
1210-35-1 | |
208.26 | |
BMVWCPGVLSILMU-UHFFFAOYSA-N | |
14589 | |
O=C1C2=CC=CC=C2CCC2=CC=CC=C12 |
C15H12O | |
MFCD00003587 | |
dibenzosuberone, dibenzocycloheptenone, dibenzosuberan-5-one, dibenzsuberone, 5-dibenzosuberone, 2,3:6,7-dibenzosuberone, 5h-dibenzo a,d cyclohepten-5-one, 10,11-dihydro, unii-8etk71th0h, dibenzo b,f cycloheptan-1-one, ccris 2780 | |
5,6-dihydrodibenzo[2,1-b |
Sécurité et manipulation
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 214-912-3
RUO – Research Use Only