missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(-)-Dibenzoyl-L-tartaric acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 438121000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(-)-Dibenzoyl-L-tartaric acid | |
97.5 | |
Authentic | |
Glass bottle | |
100g | |
dibenzoyl-l-tartaric acid, 2r,3r-2,3-bis benzoyloxy succinic acid, dibenzoyltartaric acid, --dibenzoyl-l-tartaric acid, dibenzoyl-l-tartaricacid, l-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r, unii-3q14b666vm, 2,3-bis benzoyloxy tartaric acid, o,o-dibenzoyl-+-tartaric acid | |
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O | |
358.31 | |
358.31 |
2743-38-6 | |
100.0 | |
97.5% min. (HPLC) | |
C18H14O8 | |
− 116.70 | |
YONLFQNRGZXBBF-ZIAGYGMSSA-N | |
(2R,3R)-2,3-dibenzoyloxybutanedioic acid | |
75969 | |
98% |
Chemical Identifiers
2743-38-6 | |
358.31 | |
dibenzoyl-l-tartaric acid, 2r,3r-2,3-bis benzoyloxy succinic acid, dibenzoyltartaric acid, --dibenzoyl-l-tartaric acid, dibenzoyl-l-tartaricacid, l-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r, unii-3q14b666vm, 2,3-bis benzoyloxy tartaric acid, o,o-dibenzoyl-+-tartaric acid | |
(2R,3R)-2,3-dibenzoyloxybutanedioic acid |
C18H14O8 | |
YONLFQNRGZXBBF-ZIAGYGMSSA-N | |
75969 | |
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 220-374-
RUO â Research Use Only