missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibenzoyl peroxide, 97% (dry wt.), wet with 25% water, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1317430

 View more versions of this product

Catalog No. AAL1317430


Explore available promotions

Description

Description

Benzoyl peroxide is widely utilized as a radical initiator to induce polymerizations. It finds applications for acne treatment, for bleaching flour, hair and teeth and for cross-linking polyester resins. It also has major applications in antiseptic and bleaching properties. It serves as a catalyst for polyester thermoset resins and as a hardener to start the polymerization process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoyl peroxide is widely utilized as a radical initiator to induce polymerizations. It finds applications for acne treatment, for bleaching flour, hair and teeth and for cross-linking polyester resins. It also has major applications in antiseptic and bleaching properties. It serves as a catalyst for polyester thermoset resins and as a hardener to start the polymerization process.

Solubility
Soluble in ether and chloroform. Slightlysoluble in ethanol. Insoluble in water.

Notes
Incompatible with strong oxidizer, reducing agents, acids, bases, alcohols and metals.
Specifications

Specifications

Benzoyl peroxide
(dry wt.), wet with 25% water
MFCD00003071
UN3104
14,1116
Soluble in ether and chloroform. Slightlysoluble in ethanol. Insoluble in water.
C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2
242.23
CHEBI:82405
97%
94-36-0
C14H10O4
250 g
984320
benzoyl peroxide, dibenzoyl peroxide, peroxide, dibenzoyl, benzoperoxide, benzoyl superoxide, acetoxyl, lucidol, benoxyl, panoxyl, benzol peroxide
OMPJBNCRMGITSC-UHFFFAOYSA-N
benzoyl benzenecarboperoxoate
7187
242.23

Chemical Identifiers

94-36-0
242.23
OMPJBNCRMGITSC-UHFFFAOYSA-N
7187
benzoyl benzenecarboperoxoate
C14H10O4
MFCD00003071
benzoyl peroxide, dibenzoyl peroxide, peroxide, dibenzoyl, benzoperoxide, benzoyl superoxide, acetoxyl, lucidol, benoxyl, panoxyl, benzol peroxide
CHEBI:82405
C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2
Safety and Handling

Safety and Handling

GHS H Statement
H242-H319-H317
Heating may cause a fire.
Causes serious eye irritation.
May cause an allergic skin reaction.

P210-P220-P234-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P363-P501c

H242-H317-H319-H335

DOTInformation : Hazard Class: 5.2

EINECSNumber : 202-327-6

RTECSNumber : DM8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only