Learn More
Dibenzyl malonate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1084418
Description
Dibenzyl malonate is used in the preparation of tetraethyl 3,3-bis(benzyloxycarbonyl)propylene bisphosphonate. It was also used in the preparation of benzyl umbelliferone-3-carboxylate via Knoevenagel condensation with 2,4-dihydroxybezaldehyde. It is a useful alternative to Diethyl malonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDibenzyl malonate is used in the preparation of tetraethyl 3,3-bis(benzyloxycarbonyl)propylene bisphosphonate. It was also used in the preparation of benzyl umbelliferone-3-carboxylate via Knoevenagel condensation with 2,4-dihydroxybezaldehyde. It is a useful alternative to Diethyl malonate.
Solubility
Sparingly soluble in water; miscible in all proportions with ether and alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifications
Dibenzyl malonate | |
1.137 | |
>110°C (230°F) | |
1.544 | |
50 g | |
dibenzyl malonate, propanedioic acid, bis phenylmethyl ester, 1,3-dibenzyl propanedioate, malonic acid dibenzyl ester, malonic acid dibenzyl, acmc-209d3u, diphenylmethyl propane-1,3-dioate, propanedioic acid,1,3-bis phenylmethyl ester, propanedioic acid, 1,3-bis phenylmethyl ester, s-3-amino-2-oxetanone p-toluenesulfonic acid salt boc protection form | |
RYFCSKVXWRJEOB-UHFFFAOYSA-N | |
dibenzyl propanedioate | |
84754 | |
95% |
15014-25-2 | |
188°C (0.2 mmHg) | |
C17H16O4 | |
MFCD00004779 | |
1998264 | |
Sparingly soluble in water; miscible in all proportions with ether and alcohol. | |
C1=CC=C(C=C1)COC(=O)CC(=O)OCC2=CC=CC=C2 | |
284.311 | |
284.31 |
Chemical Identifiers
15014-25-2 | |
284.311 | |
RYFCSKVXWRJEOB-UHFFFAOYSA-N | |
84754 | |
C1=CC=C(C=C1)COC(=O)CC(=O)OCC2=CC=CC=C2 |
C17H16O4 | |
MFCD00004779 | |
dibenzyl malonate, propanedioic acid, bis phenylmethyl ester, 1,3-dibenzyl propanedioate, malonic acid dibenzyl ester, malonic acid dibenzyl, acmc-209d3u, diphenylmethyl propane-1,3-dioate, propanedioic acid,1,3-bis phenylmethyl ester, propanedioic acid, 1,3-bis phenylmethyl ester, s-3-amino-2-oxetanone p-toluenesulfonic acid salt boc protection form | |
dibenzyl propanedioate |
Safety and Handling
EINECSNumber : 239-099-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only