missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibromo(1,5-cyclooctadiene)platinum(II), Pt 41.6% min, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 3928203

 View more versions of this product

Catalog No. AA3928203


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Dibromo(1,5-cyclooctadiene)platinum(II)
C8H12Br2Pt
1 g
dibromo 1,5-cyclooctadiene platinum ii, c8h12.br2pt, 1z,5z-cycloocta-1,5-diene; dibromoplatinum, 1,5-cyclooctadiene platinum ii bromide, dibromo cycloocta-1,5-diene platinum ii, 1,5-cyclooctadiene, z,z-; dibromoplatinum
HFLCCHAOKSABAA-PHFPKPIQSA-L
platinum(2+) (1Z,5Z)-cycloocta-1,5-diene dibromide
92135605
Pt 41.6% min.
12145-48-1
MFCD00058724
Hygroscopic
Insoluble in water.
[Br-].[Br-].[Pt++].C1C\C=C/CC\C=C/1
463.08
463.09
Powder

Chemical Identifiers

12145-48-1
463.08
HFLCCHAOKSABAA-PHFPKPIQSA-L
92135605
C8H12Br2Pt
MFCD00058724
dibromo 1,5-cyclooctadiene platinum ii, c8h12.br2pt, 1z,5z-cycloocta-1,5-diene; dibromoplatinum, 1,5-cyclooctadiene platinum ii bromide, dibromo cycloocta-1,5-diene platinum ii, 1,5-cyclooctadiene, z,z-; dibromoplatinum
[Br-].[Br-].[Pt++].C1C\C=C/CC\C=C/1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions