missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibucaine hydrochloride, Thermo Scientific Chemicals

Potent, long-acting local anesthetic

$108.11 - $476.63

Chemical Identifiers

CAS 61-12-1
Molecular Formula C20H30ClN3O2
Molecular Weight (g/mol) 379.929
MDL Number MFCD00012735
InChI Key IVHBBMHQKZBJEU-UHFFFAOYSA-N
Synonym dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
PubChem CID 521951
ChEBI CHEBI:59735
IUPAC Name 2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
SMILES CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6280406
View Documents
Thermo Scientific Chemicals
J6280406
5 g
Each for $108.11
 
AAJ6280414
View Documents
Thermo Scientific Chemicals
J6280414
25 g
Each for $476.63
 
Description

Description

It is applied as a potent, long-acting local anesthetic. It is used as an anaesthetic agent for spinal anaesthesia. More commonly it is a component in rectal medications used in inflammation of the anus and for the symptomatic treatment of haemorrhoids and fissures in the anus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a potent, long-acting local anesthetic. It is used as an anaesthetic agent for spinal anaesthesia. More commonly it is a component in rectal medications used in inflammation of the anus and for the symptomatic treatment of haemorrhoids and fissures in the anus.

Solubility
Freely soluble in water, alcohol, acetone, or chloroform

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

61-12-1
379.929
IVHBBMHQKZBJEU-UHFFFAOYSA-N
521951
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
C20H30ClN3O2
MFCD00012735
dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
CHEBI:59735
CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl

Specifications

61-12-1
C20H30ClN3O2
5 g
dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
IVHBBMHQKZBJEU-UHFFFAOYSA-N
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
521951
379.92
Dibucaine hydrochloride
White
MFCD00012735
14,3031
Freely soluble in water, alcohol, acetone, or chloroform
CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl
379.929
CHEBI:59735
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

EINECSNumber : 200-498-1

RTECSNumber : GD3325000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only