missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dibutyl Disulfide 98.5+%, TCI America™
Supplier: TCI America D139525ML
Specifications
Dibutyl Disulfide [for Determination of Sulfur Content by Energy Dispersive X-ray Fluorescence Metho | |
Yellow | |
C8H18S2 | |
25 mL | |
CUDSBWGCGSUXDB-UHFFFAOYSA-N | |
1-(butyldisulfanyl)butane | |
12386 | |
≥98.5% (GC) |
629-45-8 | |
226°C | |
MFCD00009467 | |
dibutyl disulfide, butyl disulfide, disulfide, dibutyl, n-butyl disulfide, di-n-butyl disulfide, dibutyl disulphide, 5,6-dithiadecane, 1-butyldisulfanyl butane, unii-z77ef35c0q, 1-butyldisulfanyl-butane | |
CCCCSSCCCC | |
178.35 | |
178.35 | |
Liquid |
Chemical Identifiers
629-45-8 | |
178.35 | |
CUDSBWGCGSUXDB-UHFFFAOYSA-N | |
12386 | |
CCCCSSCCCC |
C8H18S2 | |
MFCD00009467 | |
dibutyl disulfide, butyl disulfide, disulfide, dibutyl, n-butyl disulfide, di-n-butyl disulfide, dibutyl disulphide, 5,6-dithiadecane, 1-butyldisulfanyl butane, unii-z77ef35c0q, 1-butyldisulfanyl-butane | |
1-(butyldisulfanyl)butane |
Safety and Handling
EINECSNumber : (2)-0477
TSCA : Yes