missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyl Itaconate (stabilized with HQ) 97.0+%, TCI America™
$44.73 - $134.21
Chemical Identifiers
| CAS | 2155-60-4 |
|---|---|
| Molecular Formula | C13H22O4 |
| Molecular Weight (g/mol) | 242.315 |
| MDL Number | MFCD00027211 |
| InChI Key | OGVXYCDTRMDYOG-UHFFFAOYSA-N |
| Synonym | Itaconic Acid Dibutyl Ester |
| PubChem CID | 75080 |
| IUPAC Name | dibutyl 2-methylidenebutanedioate |
| SMILES | CCCCOC(=O)CC(=C)C(=O)OCCCC |
Chemical Identifiers
| 2155-60-4 | |
| 242.315 | |
| OGVXYCDTRMDYOG-UHFFFAOYSA-N | |
| 75080 | |
| CCCCOC(=O)CC(=C)C(=O)OCCCC |
| C13H22O4 | |
| MFCD00027211 | |
| Itaconic Acid Dibutyl Ester | |
| dibutyl 2-methylidenebutanedioate |
Specifications
| 2155-60-4 | |
| 190°C | |
| MFCD00027211 | |
| Itaconic Acid Dibutyl Ester | |
| CCCCOC(=O)CC(=C)C(=O)OCCCC | |
| 242.315 | |
| 242.32 | |
| Liquid |
| Yellow | |
| C13H22O4 | |
| 25 g | |
| OGVXYCDTRMDYOG-UHFFFAOYSA-N | |
| dibutyl 2-methylidenebutanedioate | |
| 75080 | |
| ≥97.0% (GC) | |
| Dibutyl Itaconate (stabilized with HQ) |
Safety and Handling
EINECSNumber : (2)-1130
TSCA : Yes