missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibutyl Malonate 99.0+%, TCI America™

$160.90 - $1313.14

Chemical Identifiers

CAS 1190-39-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00053795
InChI Key NFKGQHYUYGYHIS-UHFFFAOYSA-N
Synonym Malonic Acid Dibutyl Ester
PubChem CID 136925
IUPAC Name 1,3-dibutyl propanedioate
SMILES CCCCOC(=O)CC(=O)OCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M054025G
View Documents
TCI America
M054025G
25 g
Each for $160.90
 
M0540500G
View Documents
TCI America
M0540500G
500 g
Each for $1,313.14
 
Description

Description

Specifications

Chemical Identifiers

1190-39-2
216.28
NFKGQHYUYGYHIS-UHFFFAOYSA-N
136925
CCCCOC(=O)CC(=O)OCCCC
C11H20O4
MFCD00053795
Malonic Acid Dibutyl Ester
1,3-dibutyl propanedioate

Specifications

1190-39-2
251°C
MFCD00053795
Malonic Acid Dibutyl Ester
CCCCOC(=O)CC(=O)OCCCC
216.28
216.28
Liquid
Colorless
C11H20O4
25 g
NFKGQHYUYGYHIS-UHFFFAOYSA-N
1,3-dibutyl propanedioate
136925
≥99.0% (GC)
Dibutyl Malonate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'