Dibutyl Phosphate (Mono- and Di- Ester mixture), TCI America™
Supplier:  TCI America P026725G
Catalog No. P026725G
Explore available promotions
Description
Description
Specifications
Specifications
Dibutyl Phosphate (Mono- and Di- Ester mixture)
Yellow
MFCD00015245
1718
JYFHYPJRHGVZDY-UHFFFAOYSA-N
dibutoxyphosphinic acid
7881
Liquid
107-66-4
C8H19O4P
25 g
dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
CCCCOP(O)(=O)OCCCC
210.21
210.21
Chemical Identifiers
107-66-4
210.21
JYFHYPJRHGVZDY-UHFFFAOYSA-N
7881
CCCCOP(O)(=O)OCCCC
C8H19O4P
MFCD00015245
dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
dibutoxyphosphinic acid
Videos
Safety and Handling
Safety and Handling
EINECSNumber : (2)-1986
RTECSNumber : TB9605000
TSCA : Yes