missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dibutyl Phthalate 97.0+%, TCI America™
$37.52 - $41.48
Chemical Identifiers
CAS | 84-74-2 |
---|---|
Molecular Formula | C16H22O4 |
Molecular Weight (g/mol) | 278.35 |
MDL Number | MFCD00009441 |
InChI Key | DOIRQSBPFJWKBE-UHFFFAOYSA-N |
Synonym | dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db |
PubChem CID | 3026 |
ChEBI | CHEBI:34687 |
IUPAC Name | 1,2-dibutyl benzene-1,2-dicarboxylate |
SMILES | CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
P029225ML
|
TCI America
P029225ML |
25 mL |
Each for $37.52
|
|
||||||
P0292500ML
|
TCI America
P0292500ML |
500 mL |
Each for $41.48
|
|
||||||
Chemical Identifiers
84-74-2 | |
278.35 | |
DOIRQSBPFJWKBE-UHFFFAOYSA-N | |
3026 | |
1,2-dibutyl benzene-1,2-dicarboxylate |
C16H22O4 | |
MFCD00009441 | |
dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db | |
CHEBI:34687 | |
CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC |
Specifications
84-74-2 | |
Yellow | |
C16H22O4 | |
25 mL | |
dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db | |
CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC | |
278.35 | |
CHEBI:34687 | |
≥97.0% (GC) | |
Dibutyl Phthalate |
-35°C | |
340°C | |
MFCD00009441 | |
3082 | |
DOIRQSBPFJWKBE-UHFFFAOYSA-N | |
1,2-dibutyl benzene-1,2-dicarboxylate | |
3026 | |
278.35 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-1303
RTECSNumber : TI0875000
TSCA : Yes