missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibutylgermanium dichloride, 98%, Thermo Scientific™

$559.07 - $559.07

Chemical Identifiers

CAS 4593-81-1
Molecular Formula C8H18Cl2Ge
Molecular Weight (g/mol) 257.73
MDL Number MFCD00013590
InChI Key VKMPTHGEGBKPBQ-UHFFFAOYSA-N
Synonym di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4
PubChem CID 20723
IUPAC Name dibutyl(dichloro)germane
SMILES CCCC[Ge](CCCC)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC381110050
SDS View Documents
Thermo Scientific Chemicals
381110050
5 g Glass bottle
Each for $559.07
 
Description

Description

Specifications

Chemical Identifiers

4593-81-1
257.73
VKMPTHGEGBKPBQ-UHFFFAOYSA-N
20723
CCCC[Ge](CCCC)(Cl)Cl
C8H18Cl2Ge
MFCD00013590
di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4
dibutyl(dichloro)germane

Specifications

4593-81-1
1.2080g/mL
Authentic
Glass bottle
1.4720 to 1.474
MFCD00013590
1.208
VKMPTHGEGBKPBQ-UHFFFAOYSA-N
dibutyl(dichloro)germane
20723
98%
Dibutylgermanium dichloride
Colorless
242°C
97.5% min. (GC)
C8H18Cl2Ge
(CH3CH2CH2CH2)2Ge(Cl)2
5 g
di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4
CCCC[Ge](CCCC)(Cl)Cl
257.73
257.73
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-986-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only