missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibutyltin oxide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 179361000

 View more versions of this product

Catalog No. AC179361000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dibutyltin oxide
818-08-6
White
98%
C8H18OSn
MFCD00001992
04, I, 588
15, 3048
Solubility in water: soluble. Other solubilities: practically insoluble in organic solvents
CCCC[Sn](=O)CCCC
248.92
248.92
Powder or Granular Powder
98%
>300.0°C
Authentic
Plastic bottle
[CH3(CH2)3]2Sn(=O)
100 g
05,188; 09,141; 10,123; 12,160; 13,53; 15,116; 16,112
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
JGFBRKRYDCGYKD-UHFFFAOYSA-N
dibutyl(oxo)tin
61221
98%

Chemical Identifiers

818-08-6
248.92
JGFBRKRYDCGYKD-UHFFFAOYSA-N
61221
CCCC[Sn](=O)CCCC
C8H18OSn
MFCD00001992
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
dibutyl(oxo)tin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.
Toxic if swallowed.
Suspected of causing genetic defects.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Very toxic

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa

GHS Signal Word: Danger

EINECSNumber : 212-449-1

RTECSNumber : WH7175000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only