missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer, Thermo Scientific Chemicals

$222.79 - $807.16

Chemical Identifiers

CAS 50982-12-2
Molecular Formula C8H12Cl2Ru
Molecular Weight (g/mol) 280.15
MDL Number MFCD00171304
InChI Key DMRVBCXRFYZCPR-PHFPKPIQSA-L
Synonym dichloro cycloocta-1,5-diene ruthenium ii, dichloro 1,5-cyclooctadiene ruthenium ii, 1,5-cyclooctadiene, z,z-; dichlororuthenium
PubChem CID 91884784
IUPAC Name (5Z)-cycloocta-1,5-diene;dichlororuthenium
SMILES [Cl-].[Cl-].[Ru++].C1C\C=C/CC\C=C/1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1049403
View Documents
Thermo Scientific Chemicals
01049403
1 g
Each for $222.79
 
AA1049406
View Documents
Thermo Scientific Chemicals
01049406
5 g
Each for $807.16
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50982-12-2
280.15
DMRVBCXRFYZCPR-PHFPKPIQSA-L
91884784
[Cl-].[Cl-].[Ru++].C1C\C=C/CC\C=C/1
C8H12Cl2Ru
MFCD00171304
dichloro cycloocta-1,5-diene ruthenium ii, dichloro 1,5-cyclooctadiene ruthenium ii, 1,5-cyclooctadiene, z,z-; dichlororuthenium
(5Z)-cycloocta-1,5-diene;dichlororuthenium

Specifications

50982-12-2
Brown to Black
MFCD00171304
dichloro cycloocta-1,5-diene ruthenium ii, dichloro 1,5-cyclooctadiene ruthenium ii, 1,5-cyclooctadiene, z,z-; dichlororuthenium
[Cl-].[Cl-].[Ru++].C1C\C=C/CC\C=C/1
280.15
Powder
200°C (decomposition)
C8H12Cl2Ru
1 g
DMRVBCXRFYZCPR-PHFPKPIQSA-L
(5Z)-cycloocta-1,5-diene;dichlororuthenium
91884784
Dichloro(1,5-cyclooctadiene)ruthenium(II), Polymer
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only