missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dichloroacetic Acid 98.0+%, TCI America™
Supplier: TCI America D030825G
Specifications
Dichloroacetic Acid | |
1.2 (129g/L H2O soln.) | |
C2H2Cl2O2 | |
25 g | |
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid | |
C(C(=O)O)(Cl)Cl | |
128.936 | |
CHEBI:36386 | |
≥98.0% (GC,T) |
79-43-6 | |
193°C | |
MFCD00004223 | |
1764 | |
JXTHNDFMNIQAHM-UHFFFAOYSA-N | |
2,2-dichloroacetic acid | |
6597 | |
128.94 | |
Liquid |
Chemical Identifiers
79-43-6 | |
128.936 | |
JXTHNDFMNIQAHM-UHFFFAOYSA-N | |
6597 | |
2,2-dichloroacetic acid |
C2H2Cl2O2 | |
MFCD00004223 | |
dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid | |
CHEBI:36386 | |
C(C(=O)O)(Cl)Cl |
Safety and Handling
EINECSNumber : (2)-1161
RTECSNumber : AG6125000
TSCA : Yes