missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloroacetyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 113081000

 View more versions of this product

Catalog No. AC113081000


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Dichloroacetyl chloride
79-36-7
1.5300g/mL
66°C
96% min. (GC)
C2HCl3O
Cl2CHCOCl
100 mL
1.53
dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove
FBCCMZVIWNDFMO-UHFFFAOYSA-N
2,2-dichloroacetyl chloride
6593
147.39
Liquid
97%
Colorless to Yellow
107.0°C to 108.0°C
Authentic
Glass bottle
1.4600 to 1.4620
MFCD00000840
02, 204
15, 3062
Solubility in water: may decompose. Other solubilities: soluble in ether,decomposes in alcohol
C(C(=O)Cl)(Cl)Cl
147.39
CHEBI:34688
97%

Chemical Identifiers

79-36-7
147.39
FBCCMZVIWNDFMO-UHFFFAOYSA-N
6593
2,2-dichloroacetyl chloride
C2HCl3O
MFCD00000840
dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove
CHEBI:34688
C(C(=O)Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-199-9

RTECSNumber : AO6650000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only