missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichlorobis(2-ethylhexyl)silane 95.0+%, TCI America™

$187.62 - $568.79

Chemical Identifiers

CAS 1089687-03-5
Molecular Formula C16H34Cl2Si
Molecular Weight (g/mol) 325.433
MDL Number MFCD22370170
InChI Key BXDGSXMFTGZLDT-UHFFFAOYSA-N
PubChem CID 22023834
IUPAC Name dichloro-bis(2-ethylhexyl)silane
SMILES CCCCC(CC)C[Si](CC(CC)CCCC)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D44431G
View Documents
TCI America
D44431G
1 g
Each for $187.62
 
D44435G
View Documents
TCI America
D44435G
5 g
Each for $568.79
 
Description

Description

Specifications

Chemical Identifiers

1089687-03-5
325.433
BXDGSXMFTGZLDT-UHFFFAOYSA-N
dichloro-bis(2-ethylhexyl)silane
C16H34Cl2Si
MFCD22370170
22023834
CCCCC(CC)C[Si](CC(CC)CCCC)(Cl)Cl

Specifications

1089687-03-5
C16H34Cl2Si
1 g
BXDGSXMFTGZLDT-UHFFFAOYSA-N
dichloro-bis(2-ethylhexyl)silane
22023834
≥95.0% (GC)
Dichlorobis(2-ethylhexyl)silane
Yellow
MFCD22370170
2987
CCCCC(CC)C[Si](CC(CC)CCCC)(Cl)Cl
325.433
325.43
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'