missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichlorodimethylgermane, 95+%, Thermo Scientific Chemicals

$172.94 - $172.94

Chemical Identifiers

CAS 1529-48-2
Molecular Formula C2H6Cl2Ge
Molecular Weight (g/mol) 173.57
MDL Number MFCD00013588
InChI Key YQECBLVSMFAWIZ-UHFFFAOYSA-N
Synonym dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
PubChem CID 73724
IUPAC Name dichloro(dimethyl)germane
SMILES C[Ge](C)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC316500010
SDS View Documents
Thermo Scientific Chemicals
316500010
1 g Glass bottle
Each for $172.94
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1529-48-2
173.57
YQECBLVSMFAWIZ-UHFFFAOYSA-N
73724
C[Ge](C)(Cl)Cl
C2H6Cl2Ge
MFCD00013588
dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
dichloro(dimethyl)germane

Specifications

1529-48-2
Colorless
123°C
95% min
C2H6Cl2Ge
(CH3)2GeCl2
1 g
1.5
Solubility in water: reacts
C[Ge](C)(Cl)Cl
173.57
173.57
Liquid
-22°C
1.5000g/mL
32°C
Glass bottle
1.4590 to 1.461
MFCD00013588
2920
dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
YQECBLVSMFAWIZ-UHFFFAOYSA-N
dichloro(dimethyl)germane
73724
95+%
Dichlorodimethylgermane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 216-216-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only