Learn More
Dichloromethane, 99.5%, for analysis, stabilized with ethanol, Thermo Scientific Chemicals
$190.81 - $190.81
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC390700025
|
Thermo Scientific Chemicals
390700025 |
2.5 L | Glass Bottle |
Each for $190.81
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
-97.0°C | |
0.0005 meq/g max. | |
39.0°C to 40.0°C | |
Glass Bottle | |
1.4230 to 1.4250 | |
99.4% min. (GC) | |
CH2Cl2 | |
MFCD00000881 | |
0.02% max. (K.F.) | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.93 | |
Analytical |
75-09-2,64-17-5 | |
1.3250g/mL | |
2.5 L | |
Liquid | |
0.0005% max. | |
1.325 | |
CH2Cl2 | |
0.43 mPa.s (20°C) | |
15, 6135 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and dmf | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
99.5% | |
Dichloromethane |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
GHS P Statement
Do not handle until all safety precautions have been read and understood.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: W
GHS Signal Word: Warning
EINECSNumber : 200-838-9
RUO – Research Use Only