missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals

Dichloromethane, 99.9%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, methylene chloride, freon 30, methylene bichloride, methane, dichloro, methane dichloride, narkotil, methylene dichloride, solmethine, solaesthin, 2.5L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless to Yellow

Supplier:  Thermo Scientific Chemicals 326851000

 View more versions of this product

Catalog No. AC326851000



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dichloromethane
75-09-2,513-35-9
1.3250g/mL
100 mL
Liquid
0.0005 % max.
1.325
CH2Cl2
0.43 mPa.s (20°C)
01, 60
15, 6135
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
ClCCl
84.93
CHEBI:15767
99.9%
-97.0°C
Colorless to Yellow
39.0°C to 40.0°C
Authentic
Water <30ppm
99.9%
CH2Cl2
MFCD00000881
0.003% Max. (Coulometric)
01,676; 02,273; 04,337; 07,239; 13,106
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
6344
84.93
Extra Dry

Chemical Identifiers

75-09-2
84.93
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
dichloromethane
CH2Cl2
MFCD00000881
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
CHEBI:15767
ClCCl
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.

GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only