missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™
For gas chromatography ECD and FID SupraSolv™
$206.85 - $428.54
Chemical Identifiers
| CAS | 75-09-2 |
|---|---|
| Molecular Formula | CH2Cl2 |
| Molecular Weight (g/mol) | 84.93 |
| MDL Number | MFCD00000881 |
| InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
| Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
| PubChem CID | 6344 |
| ChEBI | CHEBI:15767 |
| IUPAC Name | dichloromethane |
| SMILES | ClCCl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1060541000
|
Technidata
1060541000 |
1 L | Glass Bottle |
Each for $206.85
|
|
||||
|
M1060542500
|
Technidata
1060542500 |
2.5 L | Glass Bottle |
Each for $428.54
|
|
||||
Chemical Identifiers
| 75-09-2 | |
| 84.93 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| 6344 | |
| dichloromethane |
| CH2Cl2 | |
| MFCD00000881 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| CHEBI:15767 | |
| ClCCl |
Specifications
| Neutral (at 20°C) | |
| 75-09-2 | |
| 1.33g/cm3 at 20°C | |
| 1 L | |
| Sweet | |
| Liquid | |
| CH2Cl2 | |
| 475 hPa (20°C) | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane | |
| 6344 | |
| 84.93 |
| -95°C | |
| Colorless | |
| 40°C | |
| 605°C (1121°F) | |
| Glass Bottle | |
| 1.42 | |
| MFCD00000881 | |
| 1593 | |
| Soluble in water: 20g/L at 20°C | |
| ClCCl | |
| 84.93 | |
| CHEBI:15767 | |
| Dichloromethane |
Safety and Handling
H351: Suspected of causing cancer.
Recommended Storage : 5°C to 30°C (41°F to 86°F), Tightly closed. Keep in a well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons. Protected from light.