missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloromethane, for HPLC, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 610050040

Catalog No. AC610050040



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
Specifications

Specifications

Dichloromethane
99.9%
(1 cm cell vs water), 0.014 max. at 254nm, 0.2 max. at 240nm, 1 max. at 235nm
1.3250g/mL
4 L
EcoSafPak™
2ppm max.
CH2Cl2
MFCD00000881
0.02% max.
01,676; 02,273; 04,337; 07,239; 13,106
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
ClCCl
84.93
CHEBI:15767
≥99.9%
-95°C
75-09-2,513-35-9
>120°C
39°C to 40°C
Pass Test
1.4200 - 1.4230
0.0003 meq/g max.
CH2Cl2
Vapor Pressure: 475hPa at 20°C
01, 60
15, 6135
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
6344
84.93
HPLC

Chemical Identifiers

75-09-2
84.93
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
dichloromethane
CH2Cl2
MFCD00000881
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
CHEBI:15767
ClCCl
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Accessories
Promotions

Promotions

RUO – Research Use Only