missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diclofenac sodium salt, Thermo Scientific Chemicals

Standard NSAID and cyclooxygenase (COX) inhibitor

Supplier:  Thermo Scientific Chemicals J6260914

 View more versions of this product

Catalog No. AAJ6260914


Explore available promotions

Description

Description

Oxidation of diclofenac sodium produces the metabolite 4'-hydroxy diclofenac) which demonstrates specific inhibition of Cox-2. Inhibition of Cox by diclofenac and 4'-hydroxy diclofenac suppresses prostaglandin E2 synthesis, producing anti-inflammatory and analgesic effects. Diclofenac is also shown to stabilize the native tetrameric conformation of transthyretin (TTR) fibrils, preventing the formation of insoluble amyloidogenic TTR deposits. Diclofenac Sodium is a substrate of CYP2C9. It is also used as an inhibitor of Cox-1 and Cox-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxidation of diclofenac sodium produces the metabolite 4′-hydroxy diclofenac) which demonstrates specific inhibition of Cox-2. Inhibition of Cox by diclofenac and 4′-hydroxy diclofenac suppresses prostaglandin E2 synthesis, producing anti-inflammatory and analgesic effects. Diclofenac is also shown to stabilize the native tetrameric conformation of transthyretin (TTR) fibrils, preventing the formation of insoluble amyloidogenic TTR deposits. Diclofenac Sodium is a substrate of CYP2C9. It is also used as an inhibitor of Cox-1 and Cox-2.

Solubility
Soluble in water (50 mg/ml), PBS pH 7.2 (6 mg/ml), ethanol (∼35 mg/ml), DMF (∼35 mg/ml), DMSO (∼35 mg/ml), methanol (>24 mg/ml), and acetone (6 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

Diclofenac sodium salt
283°C to 285°C
C14H10Cl2NNaO2
25 g
14,3081
Soluble in water to 50mg/ml.
C1=CC=C(C(=C1)CC(=O)[O-])NC2=C(C=CC=C2Cl)Cl.[Na+]
318.129
CHEBI:4509
Powder
15307-79-6
White
MFCD00082251
UN2811
diclofenac sodium, sodium diclofenac, diclofenac sodium salt, voltaren, solaraze, anthraxiton, prophenatin, allvoran, batafil, delimon
KPHWPUGNDIVLNH-UHFFFAOYSA-M
sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate
5018304
318.14

Chemical Identifiers

15307-79-6
318.129
KPHWPUGNDIVLNH-UHFFFAOYSA-M
5018304
sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate
C14H10Cl2NNaO2
MFCD00082251
diclofenac sodium, sodium diclofenac, diclofenac sodium salt, voltaren, solaraze, anthraxiton, prophenatin, allvoran, batafil, delimon
CHEBI:4509
C1=CC=C(C(=C1)CC(=O)[O-])NC2=C(C=CC=C2Cl)Cl.[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H361
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 239-346-4

RTECSNumber : AG6330000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only