missing translation for 'onlineSavingsMsg'
Learn More

Didecyl phthalate, 95%

CAS: 84-77-5 | C28H46O4 | 446.67 g/mol

$265.63 - $265.63

Chemical Identifiers

CAS 84-77-5
Molecular Formula C28H46O4
Molecular Weight (g/mol) 446.67
MDL Number MFCD00041915
InChI Key PGIBJVOPLXHHGS-UHFFFAOYSA-N
Synonym didecyl phthalate, decyl phthalate, di-n-decyl phthalate, vinicizer 105, phthalic acid, didecyl ester, vinysize 105, 1,2-benzenedicarboxylic acid, didecyl ester, bis n-decyl phthalate, didecyl 1,2-benzenedicarboxylate, unii-fi5fbn947z
PubChem CID 6788
ChEBI CHEBI:34676
IUPAC Name didecyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC407100250
SDS View Documents
Thermo Scientific Chemicals
407100250
25 g Glass bottle
Each for $265.63
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

84-77-5
446.67
PGIBJVOPLXHHGS-UHFFFAOYSA-N
6788
didecyl benzene-1,2-dicarboxylate
C28H46O4
MFCD00041915
didecyl phthalate, decyl phthalate, di-n-decyl phthalate, vinicizer 105, phthalic acid, didecyl ester, vinysize 105, 1,2-benzenedicarboxylic acid, didecyl ester, bis n-decyl phthalate, didecyl 1,2-benzenedicarboxylate, unii-fi5fbn947z
CHEBI:34676
CCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCC

Specifications

84-77-5
0.9700g/mL
232°C
94% min. (GC)
C28H46O4
MFCD00041915
0.97
Solubility in water: insoluble
CCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCC
446.67
CHEBI:34676
95%
Didecyl phthalate, 95%
4.0°C
261.0°C (5.0 mmHg)
Authentic
Glass bottle
1.4790 to 1.4840
25 g
didecyl phthalate, decyl phthalate, di-n-decyl phthalate, vinicizer 105, phthalic acid, didecyl ester, vinysize 105, 1,2-benzenedicarboxylic acid, didecyl ester, bis n-decyl phthalate, didecyl 1,2-benzenedicarboxylate, unii-fi5fbn947z
PGIBJVOPLXHHGS-UHFFFAOYSA-N
didecyl benzene-1,2-dicarboxylate
6788
446.66
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-561-6

RTECSNumber : TI0900000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only