missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Didecyldimethylammonium Bromide 98.0+%, TCI America™
$78.29 - $275.87
Chemical Identifiers
| CAS | 2390-68-3 |
|---|---|
| Molecular Formula | C22H48BrN |
| Molecular Weight (g/mol) | 406.537 |
| MDL Number | MFCD00012194 |
| InChI Key | UMGXUWVIJIQANV-UHFFFAOYSA-M |
| Synonym | didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide |
| PubChem CID | 16957 |
| IUPAC Name | didecyl(dimethyl)azanium;bromide |
| SMILES | CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-] |
Chemical Identifiers
| 2390-68-3 | |
| 406.537 | |
| UMGXUWVIJIQANV-UHFFFAOYSA-M | |
| 16957 | |
| CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-] |
| C22H48BrN | |
| MFCD00012194 | |
| didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide | |
| didecyl(dimethyl)azanium;bromide |
Specifications
| 2390-68-3 | |
| C22H48BrN | |
| 5 g | |
| UMGXUWVIJIQANV-UHFFFAOYSA-M | |
| didecyl(dimethyl)azanium;bromide | |
| 16957 | |
| ≥98.0% (T) | |
| Didecyldimethylammonium Bromide |
| White-Yellow | |
| MFCD00012194 | |
| didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide | |
| CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-] | |
| 406.537 | |
| 406.54 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0184&(9)-1971
TSCA : No