missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Didodecyldimethylammonium bromide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 407120250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Didodecyldimethylammonium bromide | |
3282-73-3 | |
White | |
98.5% min. (Argentometry) | |
C26H56BrN | |
MFCD00041969 | |
didodecyldimethylammonium bromide, dilauryldimethylammonium bromide, n-dodecyl-n,n-dimethyldodecan-1-aminium bromide, didodecyl dimethyl ammonium bromide, unii-7y37a25k2i, dimethyldidodecylammonium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide, didodecyl dimethyl azanium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide 1:1 | |
CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Br-] | |
462.64 | |
462.64 | |
Crystalline Powder |
99% | |
161.0°C to 164.0°C | |
Authentic | |
Plastic bottle | |
(CH3(CH2)11)2N(Br)(CH3)2 | |
25 g | |
XRWMGCFJVKDVMD-UHFFFAOYSA-M | |
didodecyl(dimethyl)azanium;bromide | |
18669 | |
99% |
Chemical Identifiers
3282-73-3 | |
462.64 | |
XRWMGCFJVKDVMD-UHFFFAOYSA-M | |
18669 | |
CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Br-] |
C26H56BrN | |
MFCD00041969 | |
didodecyldimethylammonium bromide, dilauryldimethylammonium bromide, n-dodecyl-n,n-dimethyldodecan-1-aminium bromide, didodecyl dimethyl ammonium bromide, unii-7y37a25k2i, dimethyldidodecylammonium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide, didodecyl dimethyl azanium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide 1:1 | |
didodecyl(dimethyl)azanium;bromide |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 221-923-7
TSCA : TSCA
RUO – Research Use Only