Learn More
Diethoxydimethylsilane, 97%, Thermo Scientific Chemicals
$85.98 - $288.08
Chemical Identifiers
CAS | 78-62-6 |
---|---|
MDL Number | MFCD00009068 |
InChI Key | YYLGKUPAFFKGRQ-UHFFFAOYSA-N |
Synonym | dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22 |
PubChem CID | 62322 |
IUPAC Name | diethoxy(dimethyl)silane |
SMILES | CCO[Si](C)(C)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC174631000
|
Thermo Scientific Chemicals
174631000 |
100 mL |
Each for $85.98
|
|
|||||
AC174635000
|
Thermo Scientific Chemicals
174635000 |
500 mL |
Each for $288.08
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-62-6 | |
YYLGKUPAFFKGRQ-UHFFFAOYSA-N | |
62322 | |
CCO[Si](C)(C)OCC |
MFCD00009068 | |
dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22 | |
diethoxy(dimethyl)silane |
Specifications
78-62-6 | |
Colorless | |
114.0°C | |
Authentic | |
Glass bottle | |
1.3805 to 1.3825 | |
MFCD00009068 | |
13,76 | |
dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22 | |
YYLGKUPAFFKGRQ-UHFFFAOYSA-N | |
diethoxy(dimethyl)silane | |
62322 | |
97% | |
Diethoxydimethylsilane, 97% |
-70.0°C | |
0.8600g/mL | |
11°C | |
96% min. (GC) | |
C6H16O2Si | |
(CH3)2Si(OC2H5)2 | |
100 mL | |
0.86 | |
Solubility in water: may decompose. Other solubilities: soluble in most common organic solvents | |
CCO[Si](C)(C)OCC | |
148.28 | |
148.28 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautio
GHS Signal Word: Danger
EINECSNumber : 201-127-6
RUO – Research Use Only