Learn More
Diethoxydimethylsilane, 97%, Thermo Scientific Chemicals
$54.62 - $420.03
Chemical Identifiers
CAS | 78-62-6 |
---|---|
Molecular Formula | C6H16O2Si |
Molecular Weight (g/mol) | 148.277 |
MDL Number | MFCD00009068 |
InChI Key | YYLGKUPAFFKGRQ-UHFFFAOYSA-N |
Synonym | dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22 |
PubChem CID | 62322 |
IUPAC Name | diethoxy(dimethyl)silane |
SMILES | CCO[Si](C)(C)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1472518
|
Thermo Scientific Chemicals
L1472518 |
50 g |
Each for $54.62
|
|
||||||
AAL1472530
|
Thermo Scientific Chemicals
L1472530 |
250 g |
Each for $146.26
|
|
||||||
AAL147250B
|
Thermo Scientific Chemicals
L147250B |
1000 g |
Each for $420.03
|
|
||||||
Description
Diethoxydimethylsilane is useful for blocking hydroxyl and amino groups in organic synthesis reactions. This silylating step allows subsequent reactions to be carried out which would be adversely affected by the presence of active hydrogen in the hydroxyl or amine groups. Following the reaction step, hydroxyl or amine groups blocked with diethoxydimethylsilane may be recovered by a hydrolysis procedure. It is also used for preparing hydrophobic and release materials as well as enhancing flow of powders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethoxydimethylsilane is useful for blocking hydroxyl and amino groups in organic synthesis reactions. This silylating step allows subsequent reactions to be carried out which would be adversely affected by the presence of active hydrogen in the hydroxyl or amine groups. Following the reaction step, hydroxyl or amine groups blocked with diethoxydimethylsilane may be recovered by a hydrolysis procedure. It is also used for preparing hydrophobic and release materials as well as enhancing flow of powders.
Solubility
Reacts with water.
Notes
Store under dry inert gas. It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents
Chemical Identifiers
78-62-6 | |
148.277 | |
YYLGKUPAFFKGRQ-UHFFFAOYSA-N | |
62322 | |
CCO[Si](C)(C)OCC |
C6H16O2Si | |
MFCD00009068 | |
dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22 | |
diethoxy(dimethyl)silane |
Specifications
111°C to 113°C | |
78-62-6 | |
C6H16O2Si | |
UN2380 | |
11°C (52°F) | |
Reacts with water. | |
CCO[Si](C)(C)OCC | |
148.277 | |
148.28 | |
Mild | |
Moisture sensitive | |
Diethoxydimethylsilane |
-70°C | |
50 g | |
MFCD00009068 | |
1736110 | |
dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22 | |
YYLGKUPAFFKGRQ-UHFFFAOYSA-N | |
diethoxy(dimethyl)silane | |
62322 | |
97% | |
1.382 | |
0.865 |
Safety and Handling
GHS H Statement
H225-H335
Highly flammable liquid and vapor.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLDIETHOXYSILANE
EINECSNumber : 201-127-6
RTECSNumber : VV3590000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only