missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethoxymethane, 99%, pure, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 296455000

 View more versions of this product

Catalog No. AC296455000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethoxymethane
462-95-3,128-37-0
Colorless
87.5°C
Authentic
Glass bottle
1.3720 to 1.3740
MFCD00009246
0.831
Solubility in water: 4.2%. Other solubilities: miscible with ethanol, ethyl ether, very soluble in acetone, soluble in benzene, chloroform
CCOCOCC
104.15
10024
104.15
Pure
99%
-66.5°C
0.8310g/mL
−5°C
98.5% min. (GC)
C5H12O2
CH3CH2OCH2OCH2CH3
500 mL
diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
KLKFAASOGCDTDT-UHFFFAOYSA-N
ethoxymethoxyethane
100ppm BHT max.
0.405 mPa.s (20°C)
99%
Liquid

Chemical Identifiers

462-95-3
104.15
KLKFAASOGCDTDT-UHFFFAOYSA-N
10024
CCOCOCC
C5H12O2
MFCD00009246
diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
ethoxymethoxyethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Toxic in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume

GHS Signal Word: Danger

EINECSNumber : 207-330-6

RTECSNumber : PA8500000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only