missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 2,2'-Thiodiglycolate 97.0+%, TCI America™

Supplier:  TCI America D41475G

Catalog No. D41475G


Explore available promotions

Description

Description

Specifications

Specifications

Diethyl 2,2′-Thiodiglycolate
105°C
MFCD00087932
2,2′C-Thiodiglycolic Acid Diethyl Ester, Diethyl 2,2′C-Thiodiacetate, 2,2′C-Thiodiacetic Acid Diethyl Ester
CCOC(=O)CSCC(=O)OCC
206.256
206.26
Liquid
925-47-3
C8H14O4S
5 g
TVCSSJHLVLMADJ-UHFFFAOYSA-N
ethyl 2-(2-ethoxy-2-oxoethyl)sulfanylacetate
70216
≥97.0% (GC)

Chemical Identifiers

925-47-3
206.256
TVCSSJHLVLMADJ-UHFFFAOYSA-N
70216
CCOC(=O)CSCC(=O)OCC
C8H14O4S
MFCD00087932
2,2′C-Thiodiglycolic Acid Diethyl Ester, Diethyl 2,2′C-Thiodiacetate, 2,2′C-Thiodiacetic Acid Diethyl Ester
ethyl 2-(2-ethoxy-2-oxoethyl)sulfanylacetate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions