missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl (2,4-dichlorophenyl)malonate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2633027
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl (2,4-dichlorophenyl)malonate | |
C13H14Cl2O4 | |
2.5 g | |
FIGCCTUCURBNIF-UHFFFAOYSA-N | |
diethyl 2-(2,4-dichlorophenyl)propanedioate | |
2736112 | |
95% |
111544-93-5 | |
MFCD00831103 | |
diethyl 2-2,4-dichlorophenyl malonate, diethyl 2,4-dichlorophenyl malonate, 1,3-diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenylmalonate, 2-2,4-dichloro-phenyl-malonic acid diethyl ester, propanedioic acid,2-2,4-dichlorophenyl-,1,3-diethyl ester | |
CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl | |
305.15 | |
305.16 |
Chemical Identifiers
111544-93-5 | |
305.15 | |
FIGCCTUCURBNIF-UHFFFAOYSA-N | |
2736112 | |
CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl |
C13H14Cl2O4 | |
MFCD00831103 | |
diethyl 2-2,4-dichlorophenyl malonate, diethyl 2,4-dichlorophenyl malonate, 1,3-diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2-2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenyl propanedioate, diethyl 2,4-dichlorophenylmalonate, 2-2,4-dichloro-phenyl-malonic acid diethyl ester, propanedioic acid,2-2,4-dichlorophenyl-,1,3-diethyl ester | |
diethyl 2-(2,4-dichlorophenyl)propanedioate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only