missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 3,5-Di-tert-butyl-4-hydroxybenzylphosphonate 98.0+%, TCI America™

$141.67 - $425.00

Chemical Identifiers

CAS 976-56-7
Molecular Formula C19H33O4P
Molecular Weight (g/mol) 356.44
MDL Number MFCD00227871
InChI Key GJDRKHHGPHLVNI-UHFFFAOYSA-N
Synonym 3,5-Di-tert-butyl-4-hydroxybenzylphosphonic Acid Diethyl Ester
PubChem CID 70421
IUPAC Name diethyl [(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]phosphonate
SMILES CCOP(=O)(CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D29671G
View Documents
TCI America
D29671G
1 g
Each for $141.67
Only null left
 
D29675G
View Documents
TCI America
D29675G
5 g
Each for $425.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

976-56-7
356.44
GJDRKHHGPHLVNI-UHFFFAOYSA-N
70421
CCOP(=O)(CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C)OCC
C19H33O4P
MFCD00227871
3,5-Di-tert-butyl-4-hydroxybenzylphosphonic Acid Diethyl Ester
diethyl [(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]phosphonate

Specifications

976-56-7
White
MFCD00227871
3,5-Di-tert-butyl-4-hydroxybenzylphosphonic Acid Diethyl Ester
CCOP(=O)(CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C)OCC
356.44
356.44
Crystalline Powder
122°C
C19H33O4P
1 g
GJDRKHHGPHLVNI-UHFFFAOYSA-N
diethyl [(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]phosphonate
70421
≥98.0% (HPLC)
Diethyl 3,5-Di-tert-butyl-4-hydroxybenzylphosphonate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3376

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'