missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl (3-chloropropyl)malonate, 98%, Thermo Scientific Chemicals
$62.63 - $193.58
Chemical Identifiers
CAS | 18719-43-2 |
---|---|
Molecular Formula | C10H17ClO4 |
Molecular Weight (g/mol) | 236.692 |
MDL Number | MFCD00009169 |
InChI Key | WUOPYKKJFZHTRA-UHFFFAOYSA-N |
Synonym | diethyl 3-chloropropyl malonate, diethyl 2-3-chloropropyl malonate, malonic acid, 3-chloropropyl-, diethyl ester, 2-3-chloropropyl malonic acid diethyl ester, acmc-20al9u, diethyl ?-chloropropyl malonate, chloropropylmalonic acid diethyl ester, diethyl 2-3-chloropropyl propanedioate |
PubChem CID | 140413 |
IUPAC Name | diethyl 2-(3-chloropropyl)propanedioate |
SMILES | CCOC(=O)C(CCCCl)C(=O)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1477606
|
Thermo Scientific Chemicals
L1477606 |
5 g |
Each for $62.63
|
|
||||||
AAL1477614
|
Thermo Scientific Chemicals
L1477614 |
25 g |
Each for $193.58
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18719-43-2 | |
236.692 | |
WUOPYKKJFZHTRA-UHFFFAOYSA-N | |
140413 | |
CCOC(=O)C(CCCCl)C(=O)OCC |
C10H17ClO4 | |
MFCD00009169 | |
diethyl 3-chloropropyl malonate, diethyl 2-3-chloropropyl malonate, malonic acid, 3-chloropropyl-, diethyl ester, 2-3-chloropropyl malonic acid diethyl ester, acmc-20al9u, diethyl ?-chloropropyl malonate, chloropropylmalonic acid diethyl ester, diethyl 2-3-chloropropyl propanedioate | |
diethyl 2-(3-chloropropyl)propanedioate |
Specifications
18719-43-2 | |
102°C (1 mmHg) | |
C10H17ClO4 | |
MFCD00009169 | |
1785711 | |
WUOPYKKJFZHTRA-UHFFFAOYSA-N | |
diethyl 2-(3-chloropropyl)propanedioate | |
140413 | |
98% |
1.101 | |
>110°C (230°F) | |
1.443 | |
5 g | |
diethyl 3-chloropropyl malonate, diethyl 2-3-chloropropyl malonate, malonic acid, 3-chloropropyl-, diethyl ester, 2-3-chloropropyl malonic acid diethyl ester, acmc-20al9u, diethyl ?-chloropropyl malonate, chloropropylmalonic acid diethyl ester, diethyl 2-3-chloropropyl propanedioate | |
CCOC(=O)C(CCCCl)C(=O)OCC | |
236.692 | |
236.7 | |
Diethyl (3-chloropropyl)malonate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only