missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl(3-pyridyl)borane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6422522

 View more versions of this product

Catalog No. AAH6422522


Explore available promotions

Description

Description

Reactant for: preparation of inhibitors of ataxia telangiectasia mutated and Rad3 related (ATR) protein kinase as potential anticancer agents, N-Methylation reactions of the pyridyl group, Palladium catalyzed amination reactions, intramolecular Heck reactions, Coupling reactions and Synthesis of trisubstituted pyrimidines from polyhalopyrimidines via Suzuki coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for: preparation of inhibitors of ataxia telangiectasia mutated and Rad3 related (ATR) protein kinase as potential anticancer agents, N-Methylation reactions of the pyridyl group, Palladium catalyzed amination reactions, intramolecular Heck reactions, Coupling reactions and Synthesis of trisubstituted pyrimidines from polyhalopyrimidines via Suzuki coupling.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

Diethyl(3-pyridyl)borane
172°C to 175°C (literature)
MFCD00012348
4381494
Slightly soluble in water.
CCB(CC)C1=CN=CC=C1
147.03
147.03
Solid
89878-14-8
C9H14BN
100 g
diethyl 3-pyridyl borane, 3-diethylboryl pyridine, diethyl 3-pyridinyl borane, pyridine, 3-diethylboryl, 3-diethylboranyl pyridine, diethyl-3-pyridylborane, dithyl 3-pyridyl borane, 3-pyridyldiethylborane, diethyl 3-pyridyl bo
OJKBCQOJVMAHDX-UHFFFAOYSA-N
diethyl(pyridin-3-yl)borane
642851
98%

Chemical Identifiers

89878-14-8
147.03
OJKBCQOJVMAHDX-UHFFFAOYSA-N
642851
CCB(CC)C1=CN=CC=C1
C9H14BN
MFCD00012348
diethyl 3-pyridyl borane, 3-diethylboryl pyridine, diethyl 3-pyridinyl borane, pyridine, 3-diethylboryl, 3-diethylboranyl pyridine, diethyl-3-pyridylborane, dithyl 3-pyridyl borane, 3-pyridyldiethylborane, diethyl 3-pyridyl bo
diethyl(pyridin-3-yl)borane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only