missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 4-aminobenzylphosphonate, 99%, Thermo Scientific Chemicals

$193.58 - $1709.30

Chemical Identifiers

CAS 20074-79-7
Molecular Formula C11H18NO3P
Molecular Weight (g/mol) 243.24
MDL Number MFCD00007913
InChI Key ZVAYUUUQOCPZCZ-UHFFFAOYSA-N
Synonym diethyl 4-aminobenzylphosphonate, diethyl p-aminobenzylphosphonate, 4-diethoxyphosphorylmethyl aniline, p-aminobenzylphosphonic acid diethyl ester, diethyl p-aminobenzyl phosphonate, phosphonic acid, p-aminobenzyl-, diethyl ester, diethyl 4-aminophenyl methylphosphonate, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, timtec-bb sbb000830
PubChem CID 88358
SMILES CCOP(=O)(CC1=CC=C(N)C=C1)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2227906
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
B2227906
5 g
Each for $193.58
 
AAB2227914
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
B2227914
25 g
Each for $555.16
 
AAB2227922
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
B2227922
100 g
Each for $1,709.30
 
Description

Description

Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

Solubility
Soluble in methylene chloride, acetone and benzene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

20074-79-7
243.24
ZVAYUUUQOCPZCZ-UHFFFAOYSA-N
88358
C11H18NO3P
MFCD00007913
diethyl 4-aminobenzylphosphonate, diethyl p-aminobenzylphosphonate, 4-diethoxyphosphorylmethyl aniline, p-aminobenzylphosphonic acid diethyl ester, diethyl p-aminobenzyl phosphonate, phosphonic acid, p-aminobenzyl-, diethyl ester, diethyl 4-aminophenyl methylphosphonate, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, timtec-bb sbb000830
CCOP(=O)(CC1=CC=C(N)C=C1)OCC

Specifications

20074-79-7
C11H18NO3P
5 g
diethyl 4-aminobenzylphosphonate, diethyl p-aminobenzylphosphonate, 4-diethoxyphosphorylmethyl aniline, p-aminobenzylphosphonic acid diethyl ester, diethyl p-aminobenzyl phosphonate, phosphonic acid, p-aminobenzyl-, diethyl ester, diethyl 4-aminophenyl methylphosphonate, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, timtec-bb sbb000830
ZVAYUUUQOCPZCZ-UHFFFAOYSA-N
diethyl [(4-aminophenyl)methyl]phosphonate
88358
99%
91°C to 94°C
MFCD00007913
2941613
Soluble in methylene chloride,acetone and benzene. Insoluble in water.
CCOP(=O)(CC1=CC=C(N)C=C1)OCC
243.24
243.24
Diethyl 4-aminobenzylphosphonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 243-504-8

RTECSNumber : SZ6480000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only