missing translation for 'onlineSavingsMsg'
Learn More

Diethyl adipate, 99%

CAS: 141-28-6 | C10H18O4 | 202.25 g/mol

$66.21 - $115.99

Chemical Identifiers

CAS 141-28-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009215
InChI Key VIZORQUEIQEFRT-UHFFFAOYSA-N
Synonym diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
PubChem CID 8844
ChEBI CHEBI:34697
IUPAC Name diethyl hexanedioate
SMILES CCOC(=O)CCCCC(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1147622
View Documents
Thermo Scientific Chemicals
A1147622
100 g
Each for $66.21
Only null left
 
AAA1147636
View Documents
Thermo Scientific Chemicals
A1147636
500 g
Each for $115.99
Only null left
 
Description

Description

Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

141-28-6
202.25
VIZORQUEIQEFRT-UHFFFAOYSA-N
8844
diethyl hexanedioate
C10H18O4
MFCD00009215
diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
CHEBI:34697
CCOC(=O)CCCCC(=O)OCC

Specifications

141-28-6
1.008
>110°C (230°F)
1.4275
MFCD00009215
1780035
diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
VIZORQUEIQEFRT-UHFFFAOYSA-N
diethyl hexanedioate
8844
202.25
Diethyl adipate
-20°C
250°C to 252°C
C10H18O4
CH3CH2O2C(CH2)4CO2CH2CH3
100 g
14,162
Insoluble in water.
CCOC(=O)CCCCC(=O)OCC
202.25
CHEBI:34697
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-477-0

RTECSNumber : AV1100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only