missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Adipate 99.0+%, TCI America™
$31.68 - $98.79
Chemical Identifiers
| CAS | 141-28-6 |
|---|---|
| Molecular Formula | C10H18O4 |
| Molecular Weight (g/mol) | 202.25 |
| MDL Number | MFCD00009215 |
| InChI Key | VIZORQUEIQEFRT-UHFFFAOYSA-N |
| Synonym | diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate |
| PubChem CID | 8844 |
| ChEBI | CHEBI:34697 |
| IUPAC Name | diethyl hexanedioate |
| SMILES | CCOC(=O)CCCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A016225ML
|
TCI America
A016225ML |
25 mL |
Each for $31.68
|
|
|||||
|
A0162500ML
|
TCI America
A0162500ML |
500 mL |
Each for $98.79
|
|
|||||
Chemical Identifiers
| 141-28-6 | |
| 202.25 | |
| VIZORQUEIQEFRT-UHFFFAOYSA-N | |
| 8844 | |
| diethyl hexanedioate |
| C10H18O4 | |
| MFCD00009215 | |
| diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate | |
| CHEBI:34697 | |
| CCOC(=O)CCCCC(=O)OCC |
Specifications
| 141-28-6 | |
| Colorless | |
| C10H18O4 | |
| 25 mL | |
| VIZORQUEIQEFRT-UHFFFAOYSA-N | |
| diethyl hexanedioate | |
| 8844 | |
| 202.25 | |
| Liquid |
| -20°C | |
| 251°C | |
| MFCD00009215 | |
| diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate | |
| CCOC(=O)CCCCC(=O)OCC | |
| 202.25 | |
| CHEBI:34697 | |
| ≥99.0% (GC) | |
| Diethyl Adipate |
Safety and Handling
EINECSNumber : (2)-0861&(2)-0879
RTECSNumber : AV1100000
TSCA : Yes