missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl (aminomethyl)phosphonate oxalate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 332210050

 View more versions of this product

Catalog No. AC332210050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
Specifications

Specifications

Diethyl (aminomethyl)phosphonate oxalate
117196-73-3
95%
C5H14NO3P·C2H2O4
5g
OVHZWLGNOJBTDI-UHFFFAOYSA-N
diethoxyphosphorylmethanamine;oxalic acid
2733594
95%
95%
Authentic
Glass bottle
MFCD00800255
diethyl aminomethyl phosphonate oxalate, diethoxyphosphorylmethanamine; oxalic acid, diethyl aminomethyl phosphonate oxalate salt, phosphonic acid, p-aminomethyl-, diethyl ester, ethanedioate 1:1, phosphonic acid, aminomethyl-, diethyl ester, ethanedioate 1:1, acmc-20mn2b, diethyl aminomethylphosphonate oxalate, oxalic acid diethyl aminomethylphosphonate, diethyl aminomethyl phosphonate ethanedioate
CCOP(=O)(CN)OCC.C(=O)(C(=O)O)O
257.18
257.18

Chemical Identifiers

117196-73-3
257.18
OVHZWLGNOJBTDI-UHFFFAOYSA-N
2733594
CCOP(=O)(CN)OCC.C(=O)(C(=O)O)O
C5H14NO3P·C2H2O4
MFCD00800255
diethyl aminomethyl phosphonate oxalate, diethoxyphosphorylmethanamine; oxalic acid, diethyl aminomethyl phosphonate oxalate salt, phosphonic acid, p-aminomethyl-, diethyl ester, ethanedioate 1:1, phosphonic acid, aminomethyl-, diethyl ester, ethanedioate 1:1, acmc-20mn2b, diethyl aminomethylphosphonate oxalate, oxalic acid diethyl aminomethylphosphonate, diethyl aminomethyl phosphonate ethanedioate
diethoxyphosphorylmethanamine;oxalic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only