Learn More
Diethyl benzylphosphonate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1064530
Description
Diethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Diethyl benzylphosphonate | |
1.094 | |
>110°C (230°F) | |
1.4975 | |
250 g | |
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester | |
AIPRAPZUGUTQKX-UHFFFAOYSA-N | |
diethoxyphosphorylmethylbenzene | |
14122 | |
99% |
1080-32-6 | |
106°C to 108°C (1 mmHg) | |
C11H17O3P | |
MFCD00009078 | |
2580931 | |
Insoluble in water. | |
CCOP(=O)(CC1=CC=CC=C1)OCC | |
228.228 | |
228.23 |
Chemical Identifiers
1080-32-6 | |
228.228 | |
AIPRAPZUGUTQKX-UHFFFAOYSA-N | |
14122 | |
CCOP(=O)(CC1=CC=CC=C1)OCC |
C11H17O3P | |
MFCD00009078 | |
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester | |
diethoxyphosphorylmethylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-097-4
RTECSNumber : SZ6600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only