missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Cyanomethylphosphonate, TCI America™

$33.56 - $113.71

Chemical Identifiers

CAS 2537-48-6
Molecular Formula C6H12NO3P
Molecular Weight (g/mol) 177.14
MDL Number MFCD00001893
InChI Key KWMBADTWRIGGGG-UHFFFAOYSA-N
Synonym diethyl cyanomethylphosphonate, diethyl cyanomethyl phosphonate, diethylphosphono acetonitrile, cyanomethylphosphonic acid diethyl ester, diethylphosphonoacetonitrile, diethylcyanomethylphosphonate, diethyl cyanomethanephosphonate, phosphonic acid, cyanomethyl-, diethyl ester, cyanomethyl phosphonic acid diethyl ester
PubChem CID 75676
IUPAC Name diethyl (cyanomethyl)phosphonate
SMILES CCOP(=O)(CC#N)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C14305G
View Documents
TCI America
C14305G
5 g
Each for $33.56
Only null left
 
C143025G
View Documents
TCI America
C143025G
25 g
Each for $113.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2537-48-6
177.14
KWMBADTWRIGGGG-UHFFFAOYSA-N
75676
CCOP(=O)(CC#N)OCC
C6H12NO3P
MFCD00001893
diethyl cyanomethylphosphonate, diethyl cyanomethyl phosphonate, diethylphosphono acetonitrile, cyanomethylphosphonic acid diethyl ester, diethylphosphonoacetonitrile, diethylcyanomethylphosphonate, diethyl cyanomethanephosphonate, phosphonic acid, cyanomethyl-, diethyl ester, cyanomethyl phosphonic acid diethyl ester
diethyl (cyanomethyl)phosphonate

Specifications

2537-48-6
168°C
MFCD00001893
3278
KWMBADTWRIGGGG-UHFFFAOYSA-N
diethyl (cyanomethyl)phosphonate
75676
≥98.0% (GC)
Diethyl Cyanomethylphosphonate
Yellow
C6H12NO3P
5 g
diethyl cyanomethylphosphonate, diethyl cyanomethyl phosphonate, diethylphosphono acetonitrile, cyanomethylphosphonic acid diethyl ester, diethylphosphonoacetonitrile, diethylcyanomethylphosphonate, diethyl cyanomethanephosphonate, phosphonic acid, cyanomethyl-, diethyl ester, cyanomethyl phosphonic acid diethyl ester
CCOP(=O)(CC#N)OCC
177.14
177.14
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3973

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'