missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Diethyl D-tartrate, 99%, made from unnatural tartaric acid, Thermo Scientific Chemicals

$50.61 - $50.61

Chemical Identifiers

CAS 13811-71-7
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00064451
InChI Key YSAVZVORKRDODB-WDSKDSINSA-N
Synonym --diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
PubChem CID 117410
IUPAC Name diethyl (2S,3S)-2,3-dihydroxybutanedioate
SMILES CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC200340050
SDS View Documents Promotion Details
Thermo Scientific Chemicals
200340050
5 mL Glass bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13811-71-7
206.19
YSAVZVORKRDODB-WDSKDSINSA-N
117410
CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
C8H14O6
MFCD00064451
--diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
diethyl (2S,3S)-2,3-dihydroxybutanedioate

Specifications

13811-71-7
100.0
Colorless
280.0°C
Authentic
Glass bottle
1.4450 to 1.4470
MFCD00064451
03, I, 181
1.2
− 9.00
Solubility in water: insoluble. Other solubilities: soluble in organic solvents
CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
206.19
206.19
Viscous Liquid
98.5
17.0°C
1.2000g/mL
93°C
98.5% min. (GC)
C8H14O6
[-CH(OH)CO2C2H5]2
5 mL
11,181; 13,202; 16,54
−9° (20°C neat)
--diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
YSAVZVORKRDODB-WDSKDSINSA-N
diethyl (2S,3S)-2,3-dihydroxybutanedioate
117410
99%
(-)-Diethyl D-tartrate, Made from unnatural tartaric acid
Safety and Handling

Safety and Handling

EINECSNumber : 237-458-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only