missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl dimethylaminomethylenemalonate, 94%, Thermo Scientific Chemicals

$147.25 - $568.79

Chemical Identifiers

CAS 18856-68-3
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD00052394
InChI Key XNSXXTKUKYWLAH-UHFFFAOYSA-N
Synonym diethyl dimethylaminomethylenemalonate, diethyl 2-dimethylamino methylidene malonate, diethyl 2-dimethylaminomethylidene propanedioate, diethyl 2-dimethylamino methylene malonate, propanedioic acid, dimethylamino methylene-, diethyl ester, malonic acid, dimethylamino-methyl-, diethyl ester, malonic acid, dimethylaminomethylene-, diethyl ester, 1,3-diethyl 2-dimethylamino methylidene propanedioate, acmc-1buy0
PubChem CID 140439
IUPAC Name diethyl 2-(dimethylaminomethylidene)propanedioate
SMILES CCOC(=O)C(=CN(C)C)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2076506
View Documents
Thermo Scientific Chemicals
B2076506
5 g
Each for $147.25
 
AAB2076514
View Documents
Thermo Scientific Chemicals
B2076514
25 g
Each for $568.79
 
Description

Description

Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Derivatives of this compound, diethyl ethoxymethylenemalonate is used in the synthesis of chemotherapeutic agents using 6-aminobenzothiazoles as starting material.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

18856-68-3
215.25
XNSXXTKUKYWLAH-UHFFFAOYSA-N
140439
CCOC(=O)C(=CN(C)C)C(=O)OCC
C10H17NO4
MFCD00052394
diethyl dimethylaminomethylenemalonate, diethyl 2-dimethylamino methylidene malonate, diethyl 2-dimethylaminomethylidene propanedioate, diethyl 2-dimethylamino methylene malonate, propanedioic acid, dimethylamino methylene-, diethyl ester, malonic acid, dimethylamino-methyl-, diethyl ester, malonic acid, dimethylaminomethylene-, diethyl ester, 1,3-diethyl 2-dimethylamino methylidene propanedioate, acmc-1buy0
diethyl 2-(dimethylaminomethylidene)propanedioate

Specifications

18856-68-3
C10H17NO4
5 g
Soluble in methanol.
CCOC(=O)C(=CN(C)C)C(=O)OCC
215.25
215.25
Diethyl dimethylaminomethylenemalonate
134°C to 136°C (0.5 mmHg)
MFCD00052394
diethyl dimethylaminomethylenemalonate, diethyl 2-dimethylamino methylidene malonate, diethyl 2-dimethylaminomethylidene propanedioate, diethyl 2-dimethylamino methylene malonate, propanedioic acid, dimethylamino methylene-, diethyl ester, malonic acid, dimethylamino-methyl-, diethyl ester, malonic acid, dimethylaminomethylene-, diethyl ester, 1,3-diethyl 2-dimethylamino methylidene propanedioate, acmc-1buy0
XNSXXTKUKYWLAH-UHFFFAOYSA-N
diethyl 2-(dimethylaminomethylidene)propanedioate
140439
95%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only