missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl ether, ACS reagent, anhydrous, contains BHT as inhibitor, ≥99.0%, Honeywell Riedel-de Haën™
ACS reagent, anhydrous, contains BHT as inhibitor,≥99.0%
Supplier: Honeywell Inc 6738114L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Diethyl ether | |
ACS reagent, anhydrous, contains BHT as inhibitor, ≥99.0% | |
≤100.0000% | |
34°C to 35°C | |
−45°C | |
anhydrous | |
(CH3CH2)2O | |
UN1155 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
>99% |
-116°C (literature) | |
60-29-7 | |
0.706Â g/mLÂ at 25Â °C(lit.) | |
4 L | |
Aluminum Bottle | |
C4H10O | |
MFCD00011646 | |
1696894 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12 | |
ACS Reagent |
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether
EINECSNumber : 200-467-2
Recommended Storage : Room Temp