missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl Ether, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.8% (GC), Honeywell™ Riedel-de Haën™
Puriss. p.a., contains BHT as inhibitor, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%
Supplier: Honeywell Inc 322034X25L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Diethyl Ether | |
Reag. Ph. Eur. reag. ISO | |
Liquid | |
34.6°C | |
0.365 | |
320°F | |
n20/D 1.3530 | |
C4H10O | |
MFCD00011646 | |
1696894 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12g/mol | |
ACS/puriss. p.a. |
-116°C | |
60-29-7 | |
0.706g/mL at 25°C | |
4 x 2.5 L | |
−40°C (−40°F) | |
Glass Bottle | |
0 to 0.001 % | |
(CH3CH2)2O | |
UN1155 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
≥99.8% (GC) |
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Safety and Handling
P210-P261
H224-H302-H336
DOTInformation : Class 3 - PG 1 - Diethyl ether
EINECSNumber : 200-467-2
RTECSNumber : KI5775000