missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl ethylmalonate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 114361000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl ethylmalonate | |
133-13-1 | |
1.0000g/mL | |
88°C | |
99% | |
C9H16O4 | |
C2H5O2CCH(C2H5)CO2C2H5 | |
100 g | |
1 | |
Solubility in water: immiscible | |
CCC(C(=O)OCC)C(=O)OCC | |
188.22 | |
188.22 | |
Liquid |
99% | |
Colorless | |
75°C to 77°C (5.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4150 to 1.417 | |
MFCD00009167 | |
02, 644 | |
diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate | |
VQAZCUCWHIIFGE-UHFFFAOYSA-N | |
diethyl 2-ethylpropanedioate | |
8610 | |
99% |
Chemical Identifiers
133-13-1 | |
188.22 | |
VQAZCUCWHIIFGE-UHFFFAOYSA-N | |
8610 | |
CCC(C(=O)OCC)C(=O)OCC |
C9H16O4 | |
MFCD00009167 | |
diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate | |
diethyl 2-ethylpropanedioate |
Safety and Handling
EINECSNumber : 205-093-3
TSCA : TSCA
RUO – Research Use Only