Learn More
Diethyl iodomethylphosphonate, 98+%, Thermo Scientific Chemicals
$176.97 - $2085.49
Chemical Identifiers
CAS | 10419-77-9 |
---|---|
Molecular Formula | C5H12IO3P |
Molecular Weight (g/mol) | 278.03 |
MDL Number | MFCD00041386 |
InChI Key | PNFHTBDRPOJJTQ-UHFFFAOYSA-N |
Synonym | diethyl iodomethylphosphonate, diethyl iodomethyl phosphonate, phosphonic acid, iodomethyl-, diethyl ester, acmc-20al8r, diethyliodomethylphosphonate, diethyl iodomethanephosphonate, iodomethylphosphonic acid diethyl ester, diethyl iodomethylphosphonate 5g, iodomethyl-phosphonic acid diethyl ester |
PubChem CID | 82616 |
IUPAC Name | 1-[ethoxy(iodomethyl)phosphoryl]oxyethane |
SMILES | CCOP(=O)(CI)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1369706
|
Thermo Scientific Chemicals
A1369706 |
5 g |
Each for $176.97
|
|
||||||
AAA1369714
|
Thermo Scientific Chemicals
A1369714 |
25 g |
Each for $633.68
|
|
||||||
AAA1369722
|
Thermo Scientific Chemicals
A1369722 |
100 g |
Each for $2,085.49
|
|
||||||
Description
Diethyl iodomethylphosphonate is used in the synthesis of H2N-Tyr(tBu)-vinyl sulfone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl iodomethylphosphonate is used in the synthesis of H2N-Tyr(tBu)-vinyl sulfone.
Solubility
Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
10419-77-9 | |
278.03 | |
PNFHTBDRPOJJTQ-UHFFFAOYSA-N | |
82616 | |
CCOP(=O)(CI)OCC |
C5H12IO3P | |
MFCD00041386 | |
diethyl iodomethylphosphonate, diethyl iodomethyl phosphonate, phosphonic acid, iodomethyl-, diethyl ester, acmc-20al8r, diethyliodomethylphosphonate, diethyl iodomethanephosphonate, iodomethylphosphonic acid diethyl ester, diethyl iodomethylphosphonate 5g, iodomethyl-phosphonic acid diethyl ester | |
1-[ethoxy(iodomethyl)phosphoryl]oxyethane |
Specifications
10419-77-9 | |
>110°C (230°F) | |
1.4995 | |
5 g | |
Light sensitive | |
Immiscible with water. | |
CCOP(=O)(CI)OCC | |
278.03 | |
278.03 | |
Diethyl iodomethylphosphonate |
112°C to 114°C (1 mmHg) | |
C5H12IO3P | |
MFCD00041386 | |
1812052 | |
diethyl iodomethylphosphonate, diethyl iodomethyl phosphonate, phosphonic acid, iodomethyl-, diethyl ester, acmc-20al8r, diethyliodomethylphosphonate, diethyl iodomethanephosphonate, iodomethylphosphonic acid diethyl ester, diethyl iodomethylphosphonate 5g, iodomethyl-phosphonic acid diethyl ester | |
PNFHTBDRPOJJTQ-UHFFFAOYSA-N | |
1-[ethoxy(iodomethyl)phosphoryl]oxyethane | |
82616 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 233-895-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only